Would you like to log in again?
Ward's® Genetics of Drosophila Lab Activity
Supplier: VWR
470009-212KT 282.29 USD InStock
470009-212 470009-214
Ward's® Genetics of Drosophila Lab Activity
Specimens Live Specimens Animals - Living Material
Studying the life cycle of Drosophila and performing genetic crosses just got easier with this lab activity that includes a safe way to anesthetize the fruit flies.
  • Materials for a class of 32 students working in eight lab groups or eight students working in two lab groups
  • Parental stock of three Drosophila crosses included
  • Parental generation already crossed for convenience
Students use Ward's unique Thermo-Anesthetizer, a reusable block that immobilizes fruit flies for up to 30 minutes without chemicals, allowing for close examination. In the process, students learn how to anesthetize, sex, and count flies, study their life cycle stages, and analyze the results from monohybrid, dihybrid, and sex-linked crosses.
The kit includes the parental stock of three Drosophila crosses and a wild-type culture plus materials for extracting and staining salivary gland chromosomes. Drosophila crosses will require approximately two weeks to produce offspring that can be analyzed.
Accessories information: Refill kits contain only consumable component items and do not contain additional copy masters.
Ordering information: This kit includes a coupon for redemption of perishable materials. Redeem by mail, phone, fax, or email.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty