Would you like to log in again?
Geology Squeeze Box with Screw Mechanism
Supplier: Avantor
WARD470015-586
470015-586EA 120.99 USD InStock
470015-586
Geology Squeeze Box with Screw Mechanism
Educational Classroom Kits and Activities
Calculate While You Re-Create
Add a new quantitative dimension to your tectonics demonstrations with this innovative device. When using this box, the pressure is applied with a screw mechanism that can measure equal increments of force over distance. Students can simulate folding and faulting mountain building. Developed by science teacher, Woody Williams. Includes instruction guide. Size: 11"L x 4"W x 5"H.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty