Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycogen

Supplier: ALDON CORP SE
GG0160-5G
470301-178EA 243.49 USD InStock
470301-178
Glycogen
Chemicals for Science Education
CAS Number: 9005-79-2
Formula Weight: (162.14)n
Formula: (C6H10O5)n
Density (g/mL): >1.0
Boiling Point (°C): Decomposes
Freezing Point (°C): >300
Solubility: Water and Slightly in Alcohol
Synonyms: Animal Starch
Shelf Life (months): 36
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty