Would you like to log in again?
Griffin Tubing Glass Cutter
Supplier: HUMBOLDT MFG CO
H-13520
470122-404EA 32.08 USD InStock
470122-404
Griffin Tubing Glass Cutter
Cutters
Hardened steel wheel cuts glass tubes up to 11/2" dia. Two V guides align the tubing and wheel is spring loaded to stay open.
  • Made of nickel-plated zinc-alloy die-casting
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty