Would you like to log in again?
Gymnosperm Comparison Riker Mount, Ward’s®
Supplier: Riker Mount
R149
470128-624EA 109.95 USD InStock
470128-624
Gymnosperm Comparison Riker Mount, Ward’s®
Specimens Preserved Specimens Display Specimens
Compare examples of gymnosperm families.
  • Real specimens
  • Easy to store
  • Sealed box
A great introduction and review of the gymnosperms, this display features representatives from three families including the Ginkgo (Ginkgoaceae), Juniper (Cupressaceae), and Pine (Pinaceae). Each specimen is labeled and presented in a single exhibit.
Ordering information: As these are prepared from real specimens, the product you receive may appear different from the photograph.
Delivery information: Included are specimens of the leaves or needles, male strobilus or cone, female strobilus or cone, and fruits or seeds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty