Would you like to log in again?
Half Sheep Brain, Preserved
Supplier: VWR
WARD470001-378 WARD470000-922 WARD470000-598
470001-378EA 12.99 USD InStock
470001-378 470000-922 470000-598
Half Sheep Brain, Preserved
Specimens Preserved Specimens Preserved Animal Materials
Formalin preserved sheep brain. Available in jars or pails.
  • Brain anatomy
  • Economical
Our half brains are sagittally sectioned through the median plane, allowing for structures to be easily identified. Delve into physiological studies with this sheep brain and explore neurological anatomy similar to the human brain. Sheep brains are preserved and shipped in nontoxic Wardsafe®.
Note: Ward's® preserved sheep materials is a byproduct of the food industry. Sheep are not raised specifically for dissection. These animals are recycled" for educational use."
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty