Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier® Hand-Grip Heart Rate Monitor

Supplier: Vernier
HGH-BTA
470001-872EA 128.99 USD InStock
470001-872
Vernier® Hand-Grip Heart Rate Monitor
Electrical Sockets
Ideal for continuously monitoring heart rate before, during, and after exercise.

  • Simple and easy to use
  • Hand grips do not require electrodes or clips
  • Easy to clean and share between students
  • Radios: Bluetooth and 5 kHz RF transmission
  • Battery type: CR 2025 (user-replaceable)
  • Battery lifetime: 200 h

The design of the hand-grip heart rate monitor makes it easy to monitor. Data are wirelessly transmitted to a Vernier interface using the Heart Rate Receiver. Data can also be transmitted using Bluetooth® to LabQuest 2 (may require additional hardware), iOS®, or Android™ devices without the receiver.

For educational use only.

Wireless range has RF transmission from 80 to 100 cm and bluetooth of 10 m or more unobstructed.

Certifications: It includes heart rate hand grips, polar transmitter module, heart rate receiver.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty