Would you like to log in again?
Ward's® Hardy-Weinberg Dry Erase Board
Supplier: STEPHEN GOULD CORPORATION
330888
470023-316EA 100.6 USD InStock
470023-316
Ward's® Hardy-Weinberg Dry Erase Board
Charts
Demonstrate the Hardy-Weinberg Principle Using a Real Population
Choose any of a multitude of genetic traits, such as ability to taste PTC, presence of a widow’s peak, or any other trait, and use this board to demonstrate the Hardy-Weinberg Principle. Simply collect data from your class and calculate allele frequencies on the board. The sturdy construction will provide years of classroom use. Size: 24" x 36".
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty