Would you like to log in again?
Heart, Purkinje Fibers Slide
Supplier: Avantor
WARD470182-670
470182-670EA 7.45 USD InStock
470182-670
Heart, Purkinje Fibers Slide
Slides Prepared Slides
Illustrates Mammalian Cardiac Muscle Fibers
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Demonstrating purkinje fibers, the modified cardiac muscle fibers which occur on the periphery of the cell. These fibers facilitate rapid conduction of impulses for contraction.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty