Would you like to log in again?
Heat and Temperature Set
Supplier: Avantor
WARD470152-862
470152-862EA 164.99 USD InStock
470152-862
Heat and Temperature Set
Educational Classroom Kits and Activities
Six Different Exercises Show the Effects of Heat Energy
  • Best Seller
  • Perform Many Experiments with this Set
A variety of investigations explore conduction, convection, radiation, and specific heat. The kit includes apparatus for examining thermal expansion, expansion rates, vapor pressure, gas convection, radiant heat, and specific heat to demonstrate the effects of heat energy on solids, liquids, and gases, as well as instructions for each demonstration. Smoke paper for gas convection experiment, required, is available separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty