Would you like to log in again?
Heavyweight Spill Control Pads
Supplier: ICC THE COMPLIANCE CENTER INC.
AB-YPB50H
470129-054PK 115.3 USD InStock
470129-054
Heavyweight Spill Control Pads
Spill Control Materials
Quickly Absorbs Chemicals and Liquids
When hazardous or unknown liquids spill, use these heavyweight spill pads for quick, effective response. Spill pads are yellow for quick identification in an emergency. They are made of 100% polypropylene and absorb the most aggressive and caustic fluids. Sonic–bonded “dimples” add strength and fast wicking power. Box of 50.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty