Would you like to log in again?
Vernier® High-Voltage Electrostatics Kit
Supplier: Vernier
HVEK-CRG
470017-516EA 298.99 USD InStock
470017-516
Vernier® High-Voltage Electrostatics Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Low output current makes this safe For classroom use.
  • Source with four different outputs (750, 1500, 3000, and 6000 VDC)
  • Two spheres
Take your investigation into charge even further with this accompaniment to the Vernier Charge sensor and Faraday pail. Investigate and measure the initial distribution of charge on a sphere, the transfer of charge on contact between two spheres, and charge by induction.
Electrostatics voltage source with four different outputs (750, 1500, 3000, and 6000 VDC)
Two spheres
Ground wire
A voltage terminal.
For educational use only
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty