Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High-Volume Air Sampler

Supplier: THE SCIENCE SOURCE SE
15000
470004-390EA 621.69 USD InStock
470004-390
High-Volume Air Sampler
Samplers Air Samplers
The High-Volume Air Sampler is self-operating.

  • Indoor and Outdoor Use
  • Height: 19"
  • Diameter: 10½"
  • Filter Disks Collect Unseen Particles
  • Sampler: 110 VAC

Unlike other pumps that only gather air samples, this model collects unseen airborne particles by drawing air through special filter disks. The sampler draws air at 650 L/min and can be operated both indoors and out for any required sampling period.

It operates on 110 VAC.

Ordering information: This samples includes a detachable manometer, 25 filter disks, and operating instructions with several suggestions for experiments.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty