Would you like to log in again?
HIV Simulation Lab
Supplier: Avantor
WARD470003-744
470003-744KT 70.99 USD InStock
470003-744
HIV Simulation Lab
Educational Classroom Kits and Activities
How is AIDS transmitted? Are your students listening to the message? This HIV Simulation Lab, developed by biology teacher Ruth Ann Parham, will definitely catch their attention.
Using simple pH changes and indicators, it provides students with an effective demonstration on how the AIDS virus is transmitted via body fluids.
Students trace the transmission to determine the originator of the disease. The lab can be conducted in one class period, or you can use it as the cornerstone of your AIDS study unit.
Includes materials for 60 students and a teacher’s guide with student worksheets. Grades 7–12.
Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty