Would you like to log in again?
Preserved Horsefly
Supplier: Avantor
WARD470001-308
470001-308PK 38.95 USD InStock
470001-308
Preserved Horsefly
Specimens Preserved Specimens Preserved Animal Materials
Alcohol preserved horsefly. Available in jars
  • Entomology
  • Invertebrate anatomy
Tabanus. This biting fly features slashing mouthparts. Calliphora. Size: 1/2– 3/4".
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty