Would you like to log in again?
Ward's® How Does Temperature Affect Daphnia Heart Rate? Lab Activity
Supplier: Avantor
470024-188KT 48.99 USD InStock
470024-188
Ward's® How Does Temperature Affect Daphnia Heart Rate? Lab Activity
Educational Classroom Kits and Activities
Discover how the environment affects an ectotherm's metabolism.
Teach your students important thermoregulation mechanisms used by both ectotherms and endotherms, as well as the advantages and disadvantages for each type of organism. Students will study Daphnia under the low power magnification that you provide and draw and label what they observe. Then they will monitor the Daphnia’s heart rate as they alter the temperature of the Daphnia’s environment. The kit includes enough materials for 15 setups, a teacher’s guide, and student copymaster.
Time requirement: 45 minutes.
Ordering information: Coupon included for perishable materials. Redeem by mail, fax, phone, or e-mail. Materials included: 72 microscope slides, 20 petri dishes, 1 tube petroleum jelly, 30 disposable pipets, 1 Daphnia magna culture. Materials needed but not provided: scissors, stereomicroscopes, thermometers, timer, and ice.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty