Would you like to log in again?
Pneumatic/Hydraulic Jack Set
Supplier: VWR International
PH10617A
470149-524CS 1683.05 USD InStock
470149-524
Pneumatic/Hydraulic Jack Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
This working model hydraulic/pneumatic jack shows how hydraulic or pneumatic systems can multiply forces.
  • Purpose: to Demonstrate Hydraulic and Pneumatic Systems
  • Students Explore Concepts of Force and Pressure
  • Set Can Be Used to Demonstrate Boyle's Law
  • Four Input Cylinders
  • Jack Stand Included
Four input cylinders permit varied mechanical advantage.
Students can explore force, pressure, the difference in compressibility of gases compared to liquids, the operation of a pump, and more. The set can also be used to demonstrate Boyle’s Law using air or other gases.
Ordering information: This kit includes a jack stand, plunger, 60 cc main syringe body, hose assembly with 2 one-way valves, tee and 2 quick-connect fittings, unattached tee, hose with 2 quick-connect fittings, unattached valve, hose with 1 quick-connect fitting, hose assembly with tee, and 4 smaller syringes (3 cc, 6 cc, 12 cc, 20 cc).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty