Would you like to log in again?
Ward's® Identification of Bacteria: Infusion Lab Activity
Supplier: Avantor
WARD470014-614
470014-614KT 132.79 USD InStock
470014-614
Ward's® Identification of Bacteria: Infusion Lab Activity
Educational Classroom Kits and Activities
Re-enact One of Anton van Leeuwenhoek’s Experiments
The observation of bacteria from peppercorn and mung bean infusions gives students an appreciation for the presence of bacteria in our environment, as well as an understanding of the methods used by the early microbiologists to culture and observe bacteria. They also learn about the basic differences between Gram positive and Gram negative bacteria. It includes materials for 15 setups, teacher’s guide, and student copymaster.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty