Would you like to log in again?
Ward's® Important Animal Phyla Survey Set
Supplier: VWR
WARD470000-868
470000-868KT 44.95 USD InStock
470000-868
Ward's® Important Animal Phyla Survey Set
Specimens Preserved Specimens Preserved Animal Materials
Formalin preserved.
An assortment of specimens from the animal kingdom, one from each of the ten phyla, helps students understand its wide diversity.
Contents: clam (Mollusca), earthworm (Annelida), grassfrog (Chordata), grasshopper (Arthropoda), Hydra (Coelenterata), lingula (Brachiopoda), Planaria (Platyhelminthes), roundworm (Nematoda), sponge (Porifera), and starfish (Echinodermata). The set of ten is supplied in a single pail.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty