Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Incandescent Full Hoods

Supplier: PERFECTO MANUFACTURING
77205
470106-428EA 43.59 USD InStock
470106-428
Incandescent Full Hoods
Animal Supplies Animal Cages
Affordable hoods for 10 gal. aquaria protect aquatic specimens.

  • Injection-molded high-impact polypropylene provides a durable and sturdy case for the bulbs
  • Aquarium accessories are easily accommodated due to the notched-out back strip
  • Full-length, hinged lid provides quick access to the aquarium
  • Unit operates on 120 VAC

Length: 50 cm

Certifications: CSA approved

Ordering information: Two 15 W incandescent bulbs are required but not included.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty