Would you like to log in again?
Magnetic Field Inclination/Declination Demonstration Compass
Supplier: VWR International
PH0832N
470014-316EA 70.95 USD InStock
470014-316
Magnetic Field Inclination/Declination Demonstration Compass
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Provide a concrete visual illustration of the Earth’s magnetic field, while demonstrating the “Thumb Rule” with this compass.
The Magnetic Field Inclination/Declination Demonstration Compass features a dip needle that indicates the inclination and declination of the magnetic field. This magnetic dip needle features an aluminum mounting that reduces friction for free rotation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty