Would you like to log in again?
Inertial Balance
Supplier: VWR International
PH10590A
470149-084CS 840.5 USD InStock
470149-084
Inertial Balance
Educational Materials Physics Educational Materials Mechanics Learning Activities
The oscillatory platform can carry various masses and is free to oscillate in a horizontal plane under the restoring forces of two straight-blade spring supports.
  • Economical
  • Solid Base Plate
  • Aluminum Trays
  • Instructions included
  • For Measuring Inertial Mass of Objects
It's used to measure the inertial mass of various objects.
The balance consists of separate aluminum trays securely connected by two spring blades.
Ordering information: This kit includes a base plate, a length of nylon cord, a cylindrical mass, and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty