Would you like to log in again?
Replacement Bulbs for Infrared Lamp
48069
470005-594EA 23.2 USD InStock
470005-594
Replacement Bulbs for Infrared Lamp
Lights Infrared Lights, Lamps and Bulbs
Replacement bulbs for infrared lamp and reflector.
  • 125W and 250W bulbs available
  • Medium screw base
Bulbs have medium screw base for operation on 120V, AC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty