Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Instructor’s Demonstration Tables, Phenolic Resin Top

Supplier: DIVERSIFIED WOODCRAFTS INC SE
1114K
470117-440EA 9400 USD InStock
470117-440
Instructor’s Demonstration Tables, Phenolic Resin Top
Furniture Tables and Workbenches
The instructor's desk is constructed of solid oak, oak and hardwood veneers, this unit serves as a complete demonstration center offering a spacious work surface.

  • Three-quarter inch phenolic resin top
  • Chemical resistant, earth-friendly UV finish
  • Drawers and cupboard for storage of large items

The drawers are expertly crafted from the finest materials with dovetail joinery throughout. Drawers run on epoxy-coated steel glides with nylon bearings for smooth operation.

W×D×H: 14¾×19×5½"
Small drawers W×D×H: 33¾×19×5½"
Large drawers W×D×H: 18×15×11"
Sink W×D×H: 34½×28×29¼"
Cupboard W×H: 22×32½"

Certifications: SEFA compliant and MAS Certified Green.

Ordering information: Can also be ordered without the sink, faucet, gas cock, rod sockets or support rod. Note: mirror, and mirror/marker board will not fit this unit.

Delivery information: Equipment includes: one hot and cold mixing faucet, double gas cock, GFI protected AC duplex receptacle, polypropylene trap, set of support rods and crossbar, rubber base molding and locks for cupboard and all five drawers (two small, three large).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty