Would you like to log in again?
Investigating Magnetism Kit
Supplier: Avantor
WARD470039-734
470039-734EA 215.99 USD InStock
470039-734 470328-427
Investigating Magnetism Kit
Educational Classroom Kits and Activities
Comprehensive classroom activities and lessons all in one convenient package.
  • Includes teacher and student guide
  • Developed by the Science Kit product development team
  • Grades 7–12
  • Aligned to NGSS standards (MS/HS: PS1, PS2, PS3)
Address all the facets of physical science with Investigating Physical Science Kits. These innovative packages include apparatus and activities to fully explore a key physical science topic with an entire class of 30 students. Set up multiple stations, each with its own apparatus and accompanying experiment. Students travel from station to station in groups, applying the knowledge learned at previous stations to their following activities. Each of the experiments addresses a different aspect of the same physical science concept, so upon completion students have concrete, hands-on experience and complete understanding. A detailed activity guide provides instructions for each station experiment, and an included quick-start guide simplifies initial setup. And for your convenience, a durable storage system keeps all the apparatus, guides, and materials organized for repeat use. The stackable tote can be stored compactly with all other Investigating Physical Science Kits for a complete curriculum of activities.
Actually see how magnetic fields interact with each other using the magnets and materials provided in this comprehensive investigation of magnetism. Students complete all lab activities and learn how to draw and map a magnetic field, learn what sticks to a magnet, and discover magnetic strength. A plastic sleeve contains iron filings, for easily visualized magnetic fields without the messy clean up, and a compass is used to accurately map the magnetic field around a bar magnet.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty