Would you like to log in again?
Ward's® Investigating Plant Cells Kit
Supplier: Avantor
WARD470023-808
470023-808KT 134.99 USD InStock
470023-808
Ward's® Investigating Plant Cells Kit
Educational Classroom Kits and Activities
Identify Different Types Of Cells In Vascular Plants
  • Pre And Post-Laboratory Assessments
  • Links To Real World Concepts
  • Estimated Class/Lab Time Required: Lab 1- 45-60 Minutes, Lab 2 - 20-30 Minutes
Students will prepare three microscope slides of monocot and dicot plant stems and then examine them to differentiate the xylem, phloem, and other cell structures that make up two stem specimens. They will also be able to observe the differing configurations of the two vascular tissues. It comes with two prepared reference microscope slides for comparison and identification of monocot and dicot seed structures, materials for 15 setups, a teacher’s guide, and student copy master.
Materials Included:
Monocot Stem Sections
Dicot Stem Sections
Trichrome stain
Piccolyte 2 covering media
Macerated Stem Tissue
Microscope slides
Coverslips
Alcohol Pads
Prepared Monocot and Dicot Root Microscope Slides
Prepared Monocot and Dicot Leaf Microscope Slides
Materials Needed But Not Supplied: Microscope
Ordering information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty