Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Is Albinism a Dominant or Recessive Trait? Lab Activity
Supplier: Avantor
WARD470024-130
470024-130KT 171.99 USD InStock
470024-130
Ward's® Is Albinism a Dominant or Recessive Trait? Lab Activity
Educational Classroom Kits and Activities
Explore Genetics Using Hydroponics
Students will investigate the inheritance patterns of genetic corn seeds grown hydroponically. Then by completing Punnett squares and by employing concepts in classic Mendelian genetics, your students will determine the genotypes and phenotypes of the mutation found in the offspring. In the process, they will also learn the importance of chlorophyll in green plants.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty