Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Ward's® Is There Gold in Those Gills? Lab Activity
Supplier: Ward's
WACP470180-550 WARD470180-544
470180-544PK 29.99 USD InStock
470180-544 470180-550
Ward's® Is There Gold in Those Gills? Lab Activity
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Perform Monohybrid Crosses with Vertebrates
Your students have just discovered a small population of beautiful long-finned gold zebra fish. But in order to keep the trait alive, they must first determine the genotype of these fish. The first thing your students must do is determine if the long-finned gold trait is dominant or recessive. To do this, they will cross a long-fin gold with a wildtype to determine the genotype of both strains. Then, they will raise the F1 generation until they become mature (this will take approximately three months) and cross the F1 generation to observe the F2 generation. In the process, students will also learn about the photoperiodic breeding of organisms. The kit comes with everything you need to breed enough embryos for all of your classes, teacher’s guide, and student copymaster. A stereomicroscope, required, is available separately. Note: Coupon included for perishable materials. Redeem by mail, fax, phone, or e-mail.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty