Would you like to log in again?
Karyotyping with Plain English Map of Human Chromosomes Kit
Supplier: Avantor
WARD470100-880
470100-880KT 75.99 USD InStock
470100-880
Karyotyping with Plain English Map of Human Chromosomes Kit
Charts
Why are we unique? This question and others are tackled in this fascinating kit. Students form groups of two and examine envelopes that contain genetic information for simulated reproduction. By matching up chromosomes with their partner's set and determining the genetic traits of the offspring, students learn about dominant and recessive genes, meiosis and mitosis, genetic diseases and more.
  • Grades 7 to 10
Developed by Carole MacMullan.
Delivery information: Includes: chromosome sets and a teacher's guide with worksheets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty