Would you like to log in again?
Klebsiella pneumoniae Slide
Supplier: Avantor
WARD470180-906
470180-906EA 5.95 USD InStock
470180-906
Klebsiella pneumoniae Slide
Slides Prepared Slides
Potential Agent of Pneumonia
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Rods singly or in short chains. Capsulated. Naturally occurs in the soil and as normal flora on the skin, mouth, and in the intestines. However, becomes harmful if aspirated and makes contact with the alveoli in the lungs, particularly to individuals with compromised immune systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty