Would you like to log in again?
Kyoto Kagaku® Prostate And Rectal Exam Trainer
Supplier: KYOTO KAGAKU AMERICA, INC.
M53B-C
470128-502EA 3241.29 USD InStock
470128-502
Kyoto Kagaku® Prostate And Rectal Exam Trainer
Models Human Biology Models
Interchangeable Parts Allow For Variation
  • Latex free
  • Time efficient and great for classroom environments
  • A total of nine prostate units and four rectum units
This unisex lumbar torso is designed for advanced simulation and training in differentiation of rectal diseases by palpation. High–fidelity sphincter resistance and texture of rectal wall enables extremely realistic, hands–on DRE training. Four interchangeable rectum units are included, featuring one normal rectum, one with polyps, and two with rectal cancers. Prostate unit and endocervix unit are also included, and can be felt by the student during use. Rectum is palpable up to 2.75".
Delivery information: For distribution within North America Only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty