Would you like to log in again?
Determination of Chemical Formulas Student Activity
Supplier: LAB-AIDS INC (KITS) SE
85
470007-034EA 139.65 USD InStock
470007-034
Determination of Chemical Formulas Student Activity
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Predict the Various Formulas of Compounds Formed by Their Reactions
  • Turn abstract theory into realism
  • Gain and understanding of percentage composition and molecular weight
  • Requires one 50 minute class period
Students predict the various formulas of compounds formed by their reactions. Strengthens lab procedure skills and observation abilities, and promotes an understanding of percentage composition and molecular weight.
Ordering information: Materials include Chemplates®, Lead nitrate solution, Sodium oxalate solution, Potassium iodide solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty