Would you like to log in again?
Laboratory Engraver
Supplier: ROBERT BOSCH TOOL CORPORATION
242-9924 290-01 290-01
470007-778EA 19.2 USD InStock
470007-778 470200-528 470007-776
Laboratory Engraver
Pens Stylus and Scribers
Protect your valuable microscopes, laboratory equipment, and apparatus with this easy-to-use engraver.It permanently marks on steel, plastic, copper, brass, wood, and glass. It handles like a pencil and has an easily replaceable carbide point.
The high speed, adjustable reciprocating motor delivers 7200 strokes per minute and is designed for trouble free, heavy duty use. Complete with cord and plug for operation on 110 V, ac.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty