Would you like to log in again?
Ward's® Live Ladybug Adults
Supplier: VWR
Ward's®
WACP470188-914 WARD470324-902 WACP470189-292
470188-914EA 16.6 USD InStock
CA470189-292 470188-914 CA470188-914 470189-292 470324-902
Ward's® Live Ladybug Adults
Specimens Live Specimens Animals - Living Material
Hippodamia convergens.
  • Care instructions included
Adult Ladybugs will rely on you to provide their food. Simply soak one or two raisins in water for about 5 minutes and then blot them on a paper towel. Cut them in half and drop them in their habitat. Repeat once the raisins have disappeared and be sure that your ladybugs are always supplied with water.
If you have any plants outdoors that may be bothered by aphids, you might drop a few aphid-infested leaves in the habitat as well. Ladybugs love to eat aphids!
Ordering information: This product is not available in Canada. This item contains live or perishable materials.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty