Would you like to log in again?
LaMotte® Electronic Soil Outfit
Supplier: LA MOTTE CHEMICAL PROD CO SE
LaMotte®
1985-06 R-1985-04
470021-128EA 3998.99 USD InStock
470021-128 470021-130
LaMotte® Electronic Soil Outfit
Standards Environmental Analysis Standards
Includes Colorimeter, TDS, and pH Meters
  • Quickly provides accurate results
  • Digital colorimeter measures exact % transmittance
  • Tests water and soil
The simplified test procedures included with this outfit provide 20, 50, or 100 tests for each soil nutrient, with accurate results in minutes using easy-to-prepare soil extracts. A digital colorimeter will perform 30 different tests on water and soil and then instantly analyze color reactions with a percent transmittance readout, eliminating the guesswork of visual color comparisons. Plus, special calibration charts convert readings to ppm or lbs./acre for each nutrient. Test factors include calcium, chloride, copper, iron, manganese, magnesium, nitrogen (three forms), pH (0–14), phosphorus, potassium, dissolved salts, sulfates, and zinc. The self-contained lab also includes a conductivity/dissolved solids meter and pH meter, as well as the digital colorimeter. Contact WARD’S for a full list of available test factors. Size: 22"L x 19"W x 8"H; weight: 32 lbs. Note: Contains cyanide solution and barium compounds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty