Would you like to log in again?
Ward's® Landfills and the Environment Lab Activity
Supplier: Avantor
WARD470015-802
470015-802KT 161.99 USD InStock
470015-802
Ward's® Landfills and the Environment Lab Activity
Educational Classroom Kits and Activities
Students Work with Miniature Landfills in Hands-On Investigation
  • Create outdoor landfills
  • Sample and observe microorganisms
  • includes a teacher's guide and activity manual
Determine which solid waste materials are biodegradable, identify types of bacteria present in different kinds of soil, and test the microbial degrading process by observing the landfills you create with the materials included in this lab activity. These investigations provide data that students can use to define the biodegradability of materials, as well as determine the real function of a landfill. You will get enough materials for three landfills, a list of additional outdoor activities, a teacher’s guide, and student copymasters. Time requirements include Investigation #1 (8 weeks), Investigation #2 (4-6 weeks), Investigation #3 (5 days), and Investigation #4 (2-3 weeks).
Ordering information: Lab includes Simulated landfills, Cups with lids, Plastic (bag) liners, Small self-sealing bags, Starch agar, Nutrient agar, Petri dishes, Microscope slides, Crystal violet stain, Gloves, Magnifying glasses, Sand, Topsoil, Biodegradable plastic, Sheet of plastic, Spool of natural fiber, and Spool of synthetic fiber. Scissors, Spray bottle, Bunsen burner, and Hot plate are needed but not provided.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty