Would you like to log in again?
Light Combiner Demonstration
Supplier: Avantor
WARD470006-468
470006-468KT 65.99 USD InStock
470006-468
Light Combiner Demonstration
Educational Classroom Kits and Activities
The Light Combiner is a simple yet dramatic way to demonstrate the additive color process.
  • Developed by Joe Rich
By placing the Light Combiner on an overhead projector and adjusting the focus, the primary light colors of red, green and blue are combined to produce the secondary light colors of yellow, cyan and magenta and white light. In addition to the instructions, this booklet includes a reproducible worksheet for students to record their observations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty