Would you like to log in again?
Litmus Milk Broth
Supplier: Avantor
WARD470180-732
470180-732PK 34.5 USD InStock
470180-732
Litmus Milk Broth
Media for Microbiology Dehydrated Media for Microbiology
For demonstration of acid production, coagulation, or proteolysis of casein in culture identification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty