Would you like to log in again?
Ward's® Luster Collection
Supplier: VWR
WARD470025-260
470025-260EA 34.79 USD InStock
470025-260
Ward's® Luster Collection
Specimens Mineral Specimens Mineral Collections
Examples of Different Types of Surface Appearance
The minerals in this collection are each perfect examples of one type of luster. The set is housed in a compartmented collection box, and an identification list is included.Contents: 12 numbered samples, 1×2" Minerals: Metallic Luster: pyrite Adamantine Luster: sphalerite Vitreous Luster: quartz Resinous Luster: sulfur Greasy Luster: nepheline Pearly Luster: talcSilky Luster: satin spar Splendent Luster: micaceous hematite Shining Luster: calcite Glistening Luster: prochlorite Submetallic Luster: ilmenite Dull Luster: chalk
Caution: Due to the nature of this product it may contain asbestos.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty