Would you like to log in again?
Machine-Made Neoprene Tubing
Supplier: KISMET RUBBER PRODUCTS CORP.
5/16IN OD 51225K22
470189-076FT 1.4 USD InStock
470189-076 470189-078
Machine-Made Neoprene Tubing
Tubing Transfer Tubing
A medium wall tubing excellent for student use and other temporary assemblies to be used no longer than a year. Made from synthetic rubber and available in black or red.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty