Would you like to log in again?
MagLev Train Kit
Supplier: Avantor
WARD470003-054
470003-054EA 230.59 USD InStock
470003-054
MagLev Train Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Step Into the future with this revolutionary kit.
  • Aligned with National Standards
  • Includes two 4' long tracks and materials to create four vehicles
Developed through a partnership with the Department of Energy and Brookhaven National Laboratory, this exciting activity guides students through the construction of a magnetic levitation train. Using the activity instructions and their understanding of physics and magnetism, students observe the conditions which create the most effective train. The size of each train can be customized for both racing and research; students can also vary the load to determine its effect on the train’s performance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty