Would you like to log in again?
Marine Rotifers
Supplier: VWR
470180-252EA 18 USD InStock
470180-252
Marine Rotifers
Specimens Live Specimens Aquaria - Living Material
Brachionus sp. Examine these typical marine rotifers for a general study of their structure and feeding habits.The recommended food for marine rotifers is Dunaliella. Culture instructions are included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty