Would you like to log in again?
SP Wilmad-LabGlass Measuring Pipettes, SP Industries
Supplier: SP Industries
C-7065-5
470123-206EA 15.8 USD InStock
470123-206
SP Wilmad-LabGlass Measuring Pipettes, SP Industries
Pipettes Measuring (Mohr) Pipettes
Made of borosilicate glass.
  • Permanent color scale
  • Calibrated to deliver
  • Sizes are color-coded
The top of 5 ml and larger pipette is constricted, Mohr type.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty