Would you like to log in again?
Meissner’s Corpuscle, (Primate)
Supplier: Avantor
WARD470177-658
470177-658EA 8.95 USD InStock
470177-658
Meissner’s Corpuscle, (Primate)
Slides Prepared Slides
Mechanoreceptors that sense light touch
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Primate. Shows small encapsulated sensory receptors found in the dermis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty