Would you like to log in again?
Magnetizing and Demagnetizing Coil
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
PH0804B
470121-556EA 134.45 USD InStock
470121-556
Magnetizing and Demagnetizing Coil
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Bring older, weaker magnets back to life with this magnetizer.
  • Coil allows magnets to be either remagnetized or demagnetized
A 25cm long coil allows magnets up to 4.5cm in diameter to be either remagnetized or demagnetized. The apparatus has 960 turns and the wire has a diameter of 1.22 mm. This coil serves as a useful way to demonstrate how a magnet can be made or destroyed. Two banana plug jacks allow for an AC/DC power supply to be attached, and an on/off switch is clearly visible.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty