Would you like to log in again?
Micro Crucible Holder for Laboratory Micro Burner
Supplier: GLOBAL MARKET CONNECTIONS, LTD
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MCH-2001
470150-948CS 108.1 USD InStock
470150-948
Micro Crucible Holder for Laboratory Micro Burner
Heaters Gas Burners
The three fused rings of this crucible holder fit securely into the tripod stand's grooves to hold small crucibles.
The inner diameter of each ring measures 25 mm.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty