Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Microchemistry Thermometers
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
4-428
470104-652PK 42 USD InStock
470104-652
Microchemistry Thermometers
Thermometers Analog Thermometers
Designed specifically for microchemistry experiments, these thermometers are small enough to easily fit in the smallest microplate well, offering students an easy way to monitor temperature.
Temperature range: 0–100°C.
Package of 12.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty