Would you like to log in again?
Mimicry In Lepidoptera Riker Mount
Supplier: WHITE OWL ENTERPRISES
R370
470028-082EA 66.65 USD InStock
470028-082
Mimicry In Lepidoptera Riker Mount
Specimens Preserved Specimens Display Specimens
Teach students they can’t believe everything they see.
Using two species of butterflies, this display illustrates how one organism adopts, or mimics, the appearance of another organism that is harmful or distasteful in order to be ignored by predators. Species of lepidoptera may vary; the Monarch and Viceroy, pictured, are the most commonly used examples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty