Would you like to log in again?
Mini Demo Electric Motor
Supplier: VWR International
PH1238
470014-344EA 51.15 USD InStock
470014-344
Mini Demo Electric Motor
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Economical and effective.
  • Simplest DC electric motor model available to education
This unit features a two pole armature wound with enameled copper wire. A permanent magnetic field is produced by a removable bar magnet. A disc type commutator is incorporated and external connection to the phosphor-bronze brushes is through of a pair of 4 mm sockets. Unit operates on 6–8 volts DC. required but not included is a power supply and connecting wires.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty