Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Mitochondrion Model
Supplier: THE SCIENCE SOURCE SE
B117
470004-510EA 1037.95 USD InStock
470004-510
Mitochondrion Model
Models Botany Models
Includes A Diagram Of The Krebs Cycle
The mitochondrion, the so-called “powerhouse” of cells, is re-created in a superb, highly detailed model showing the double-wall membrane, cristae, subunits, granules localized on the inner wall, and the matrix material filling the body. The structure is shown in a longitudinal section enlarged 190,000X and in cross section enlarged 1,000,000X. The one-piece model is mounted on a base featuring a diagram of the Krebs cycle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty